VWR® Maximum, Cut Resistant Gloves
Leverandør: VWR Collection
112-5691EA 915 NOK
112-5691 112-5692 112-5693 112-5694 112-5695 112-5696
VWR® Maximum, Cut Resistant Gloves
Hansker
This VWR® glove with polyurethane coating provides superior cut protection for applications that include a high risk of cuts.
  • Nylon/spandex/polyester blend composition
  • Polyurethane (PU) coating on palm and fingertips
  • Grey knitted glove with grey coating
Ideal for handling items or tools with a high risk of cuts, including steel or glass handling, assembly, maintenance, and janitorial applications.
Sertifiseringer: Meets requirements for EN 388:2016 (4X42E). Certified by OKEO-TEX meeting Standard 100 certifying that no harmful substances are used inMore Product Information
Bestill nå
SPESIFIKASJONER
Ambidekstral/håndspesifikk Håndspesifikk
Environmentally Preferable
Engangs/flergangs gjenbrukbar
Latex/latexfri Latex-fri
Finish PU palm coating
Måler 13
Cuff style Strikket håndledd
EN norm EN 388:2016 (4X42E)