VWR®, Precision Grip Gloves with Dots
Leverandør: VWR Collection
112-5730EA 560 NOK
112-5730 112-5731 112-5732 112-5733 112-5734 112-5729
VWR®, Precision Grip Gloves with Dots
Hansker
This VWR® precision grip glove with grip dots and nitrile coating provides basic cut protection and enhanced grip.
  • ANSI A1 Cut Protection (Level A)
  • Grip dots on the palm for enhanced grip
  • Nylon composition
  • Nitrile (NBR) coating on palm and fingertips
  • Grey knitted glove with black coating
Ideal for handling, picking, packaging, automotive, or situations that require dexterity in a dry and/or dirty environment.
Sertifiseringer: Meets requirements for ANSI A1 Cut Resistance. EN 388:2016 (4131X)
Certified by OKEO-TEX meeting Standard 100 certifying thatMore Product Information