VWR® Cleanroom Gloves, Latex, Sterile, for Class 10 / ISO 4
Leverandør: VWR Collection
112-6192EA 4900 NOK
112-6192 112-6193 112-6194 112-6198 112-6195 112-6196 112-6197
VWR® Cleanroom Gloves, Latex, Sterile, for Class 10 / ISO 4
Hansker
VWR® cleanroom latex gloves provides reliable, durable and comfortable hand protection during demanding cleanroom applications.
  • Beaded cuff
  • Natural rubber latex
  • Low particle count
  • Silicone free
  • Sterile 10⁻⁶ SAL
Sertifiseringer: EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016; CAT III; AQL 1.5
Forpakning: Each pair is double packaged in a wallet within a pouch, 20 pouches per bag, 10 bags per case (200 pairs).