VWR® Cleanroom Gloves, Nitrile, Solid White, Tacky, Class 10 / ISO 4
Leverandør: VWR Collection
112-6205EA 3575 NOK
112-6205 112-6206 112-6207 112-6208 112-6209 112-6210
VWR® Cleanroom Gloves, Nitrile, Solid White, Tacky, Class 10 / ISO 4
Hansker
VWR® cleanroom nitrile gloves are manufactured from a synthetic copolymer of acrylonitrile, butadiene and methylacrylic acid, that provides reliable, durable and comfortable hand protection during demanding cleanroom applications.
  • Beaded cuff
  • Nitrile
  • Silicone-free
  • Low particle count
Sertifiseringer: N ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016; CAT III; AQL 1.5
Forpakning: 100 gloves per double bag, 10 bags per case.