VWR® Cleanroom Gloves, Latex, Non-Sterile, Class 10/ISO 4
Leverandør: VWR Collection
112-6290EA 3025 NOK
112-6290 112-6291 112-6292 112-6293
VWR® Cleanroom Gloves, Latex, Non-Sterile, Class 10/ISO 4
Hansker
VWR® Cleanroom Latex gloves provides reliable, durable, and comfortable hand protection during demanding cleanroom applications.
  • Beaded cuff
  • Natural rubber latex
  • Low particle count
  • Silicone-free
Sertifiseringer: EN ISO 374-1 Type B (KPT), EN ISO 374-5; CAT III; AQL 1.5.
Forpakning: Gloves are packaged 100 per double bag, 10 bags per case.