Pleated masks with ties, non sterile, HM4, PUREZERO®
Leverandør: Halyard
CLN1341TB
112-6508EA 2050 NOK
112-6508
Pleated masks with ties, non sterile, HM4, PUREZERO®
Masker Munnbind, renrom
The HM4 Pleated Mask with Ties is a three-layer mask with long ties that provides high particle, bacterial, and viral filtration efficiency. This mask is for industrial and cleanroom use. The mask construction consists of low-linting materials with a SO SOFT inner layer, a polyester foam nose band for added comfort, and anti-fog performance.
  • Three-layer mask
  • Long ties for easy donning
  • Designed for industrial and cleanroom use
  • Recommended for ISO Class 4 or higher and Grade B/C/D cleanrooms
  • Exclusive SO SOFT biocomponent technology inner lining for comfort