Organic reference standard, Brilliant Black BN, analytical standard, Supelco®
Leverandør: Merck
11220-25MG
11220-25MGEA 1625 NOK
11220-25MG
Organic reference standard, Brilliant Black BN, analytical standard, Supelco®
Standarder Organiske standarder
Brilliant Black BN (BB) is a azo dye which is basically a tetra sodium salt. It is a synthetic dye mainly used as food additive. It is used as colouring agent to enhance the appearance. It is usually used in jellies, brown sauces and cake mixes.
  • Used as colourant during quantification of synthetic colourants in fish roe and caviar for effective isolation using HPLC methods
Forpakning: Bottomless glass bottle. Contents are inside inserted fused cone.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=76ebe45f0c42596b206f38349ad347586bfc6f76&vid=76ebe45f0c42596b206f38349ad347586bfc6f76&prid=web&clid=SAPDAM