Fall protection kit for work positioning, Titan™
Leverandør: HONEYWELL SAFETY
1011899
113-1271EA 4450 NOK
113-1271
Fall protection kit for work positioning, Titan™
Fallvern Sett for fallbeskyttelse
Fall protection kit contains:
  • Titan™ 2-point harness with work positioning belt, front and rear anchorage
  • Titan™ CMS arrester with 2,5 m webbing lifeline
  • Adjustable work positioning lanyard with 2 twist lock karabiners