Safety harness, Revolution Comfort R2
Leverandør: HONEYWELL SAFETY
1014237 1014236
113-1272EA 4950 NOK
113-1272 139-0239
Safety harness, Revolution Comfort R2
Fallvern Fallsele
Two-point safety harness for use with fall protection equipment and a suitable anchorage point. The webbing is made from DuraFlex®, a highly elastic stretch fabric. Thanks to its unique technology and versatility, the harness is ideally suited for scaffolding, steel and mast construction applications, as well as for general use and maintenance.
  • Safety loop at the back (D-ring) and the front (webbing loops)
  • PivotLink connection at waist height for increased mobility and comfort thanks to optimum ergonomics when bending and stretching
  • Single-handed