Harnesses, Ropax
Leverandør: HONEYWELL SAFETY
1014435
113-1274EA 8750 NOK
113-1274
Harnesses, Ropax
Fallvern Fallsele
This harness is ideal for industrial rope access, rescue and special forces. It can be used in conjunction with a climber's seat and an ascender.
  • Black polyester webbing and stainless steel components offer optimum durability, chemical and abrasion resistance
  • Triangular Suspension position splits the forces between the leg straps and the waist belt across four points of pressure
  • Detachable webbing sling on the chest straps positions the ascender correctly in a vertical position to facilitate ascension
EN 358, EN 813, EN 361
Sertifiseringer: EN 358,More Product Information