Self-retracting lifelines, Miller® Falcon™
Leverandør: HONEYWELL SAFETY
1016796
113-1279EA 9025 NOK
113-1279
Self-retracting lifelines, Miller® Falcon™
Fallvern Fallvern
This self-retracting lifeline is the ideal fall protection solution for construction, industry, warehouses, utilities, mining and confined space applications.
  • Quick-activating braking mechanism stops a fall within centimetres
  • Lightweight
  • No annual factory re-certification required: Reduced down time, lower cost of ownership and increased productivity
  • High-impact resistant polyamide housing, polyester webbing
Sertifiseringer: EN 360
Bestill nå
SPESIFIKASJONER
Modell Falcon™
Environmentally Preferable
Kabeltype Polyester webbing
Type kobling Snap hook, aluminium
Bruddstyrke > 1500 daN
Driftstemperatur −30...+50 °C
Hus, B × D × H 540×145×70 mm