Munnbind for renrom, BioClean 100™, PP
Leverandør: Nitritex
MTA210-1
113-2250EA 12875 NOK
113-2250
Munnbind for renrom, BioClean 100™, PP
Masker Munnbind, renrom
Providing high particle and bacterial filtration efficiency, the latex-free and low-linting BioClean MTA™ sterile looped facemask ensures protection, combined with comfort and exceptional breathability.
  • Low particle shedding with excellent breathability
  • Three ply ultrasonic sealed construction: Low-linting hydrophilic inner layer, non-woven meltblown polyester filter layer, non-woven polyester outer facing
  • PP-ties and single steel PP coated noseband
  • Average particle count: 19/minute (particle size: 0,2 to 5,0 µm)
  • Particle filtration efficiency (PFE): 98% &