VWR®, Cleanroom Sleeve Protectors
Leverandør: VWR Collection
113-7887EA 4750 NOK
113-7887
VWR®, Cleanroom Sleeve Protectors
Engangsplagg
Tapered sleeve protectors made from breathable SMS. Ideal for use in cleanroom and ESD environments.
  • Elastic at both ends
  • Folded seams
  • Vacuum packed
Weight: 60 g.
Level I acc. IEST-RP-CC03.3. Suitable for use in ISO Class 5 + 6 cleanroom environments.
Sertifiseringer: Type 5/6; Cat. III.
Forpakning: Individually double-bagged with lining.