VWR®, Beard Cover, PP
Leverandør: VWR Collection
113-7920EA 875 NOK
113-7920
VWR®, Beard Cover, PP
Engangsplagg
Basic beard cover made from non woven PP. For non critical applications.
  • Elasticated around the neck
  • Breathable, soft material
  • Latex-free
Size: 46 cm
Weight: 11 g/m²