VWR®, Wall Mount for Four Glove or Clothing Boxes
Leverandør: VWR Collection
113-8040EA 5025 NOK
113-8040
VWR®, Wall Mount for Four Glove or Clothing Boxes
PPE-dispensere
The adjustable holding points provide true flexibility to hold up to four different boxed protective products, and other lab essentials. Ideal for different boxed sizes of gloves, face masks, ear plugs or shoe covers.
  • Four holding positions can hold up to four boxes in one holder
  • Wall- or door-mountable, mounting parts included
  • No restrictions regarding box size: Open ended holding points are able to hold longer and wider boxes
  • Easy to clean, with low profile