Safety spectacles dispenser
Leverandør: Palbam
GD-0302
113-8174EA 7750 NOK
113-8174
Safety spectacles dispenser
PPE-dispensere
This dispenser is suitable for use with safety spectacles in the cleanroom industries, such as pharmaceutical, biotechnology, medical device, microelectronics and nanotechnology.
  • High-quality design
  • Four holes for stable wall mounting
  • Clear front panel for higher visibility
  • Protection against splashes and impacts
  • Stainless steel 304, electropolished finish
Sertifiseringer: ISO certified
Leveringsinformasjon: Typically available from stock for direct shipment from the supplier.