Visitor coats, PP
Leverandør: VWR Collection
113-9261EA 0 NOK
113-9261 113-9260 113-8328 113-8329 113-8330 113-9262
Visitor coats, PP
Engangsplagg
These disposable coats made of PP are ideal for visitors in non hazardous environments.
  • With collar
  • Press-stud fastening (5 press-studs)
  • No pockets
Weight: 35 g/m²
Colour: White
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
lukking type Med trykknapper
Lommer uten lommer
Kjønn Unisex
Antrekksapplikasjoner General Purpose,Laboratory,Food
Weight 35 g/m²