Fall protection harnesses, Miller H-Design® BodyFit Auto 2LO
Leverandør: HONEYWELL SAFETY
1033536 1033537 1033538
113-8654EA 7150 NOK
113-8654 113-8655 113-8656
Fall protection harnesses, Miller H-Design® BodyFit Auto 2LO
Fallvern Fallsele
Miller H-Design® BodyFit harness with two sternal webbing loops and automatic buckles on chest and legs straps and DuraFlex Stretch webbing. The Miller H-Design® full body harness is dedicated to electricity, telecommunications and general industry (maintenance) sectors, providing more comfort, freedom of movement and safety to workers thanks to its patented vertically adjustable belt and breathable padding.
  • Straps are made from Duraflex Stretch and black webbing (PES with PTFE treatment)
  • Straps are water repellent and anti-stain treated
  • Two sternal