Self-retracting lifeline, Miller® TurboLite™ edge
Leverandør: HONEYWELL SAFETY
1033482
113-8668EA 3100 NOK
113-8668
Self-retracting lifeline, Miller® TurboLite™ edge
Fallvern Fallvern
A lightweight edge-tested self-retracting lifeline for all work situations, including those that require the possibility to connect at foot level when working on the edge. Ideal for construction (scaffold erecting/dismantling, steel erecting), general industries (maintenance) and utilities (wind, power and telecom; work on pylons), oil and gas.
  • Adapted to all fall factors: FF0, FF1 and FF2
  • Edge-tested - reinforced webbing makes horizontal use possible without restrictions
  • Tested at 140 kg (even in FF2 'foot level' anchorage)
  • Lightweight SRL: 1 kg (SRL with