Bouffant cap, BioClean™ BBC
Leverandør: Nitritex
BBC-W
113-8790EA 1825 NOK
113-8790
Bouffant cap, BioClean™ BBC
Engangsplagg
These lightweight cleanroom bouffant caps are made of non woven PP.
  • Hypoallergenic, soft fabric
  • Elasticated double stitching
  • Latex-free
  • Compatible with a variety of environments
Density: 10 g/m²