Hygiene mask, reusable, 2-ply
Leverandør: ALSICO HIGHTECH
1031328
113-9935EA 0 NOK
113-9935
Hygiene mask, reusable, 2-ply
Masker Munnbind
Reusable, 2-ply face masks are not designed for medical use. Masks are not tested on the permeability of viruses and bacteria.
  • Washable, autoclavable (120 °C, 20 min)
  • High filtration, high wearing comfort, low linting
  • Equipped with two binding tape fasteners
  • Cleanroom Class: ISO 3-8 (EU GMP A-C)
99% polyester, 1% carbon construction. Masks provide 98,9% filter efficiency against 0,5 µm particles, 97,9% filter efficiency against 0,3 µm particles.
Colour: White.