Dysprosium trichloride hydrate ≥99.99% (REO, rare earth oxide basis)

Leverandør: Thermo Fisher Scientific
Warning

11317.06 11317.14 11317.22
11317.06EA 1450 NOK
11317.06 11317.14 11317.22
Dysprosium trichloride hydrate ≥99.99% (REO, rare earth oxide basis)
Dysprosium trichloride hydrate

Purity: 99.99% Molecular Formula: DyCl3.xH2O Formula Weight: 268.86(anhy) Physical Form: Wet Crystalline Aggregates Sensitivity: Hygroscopic

Formel: Cl₃Dy·xH₂O
Smeltepunkt: 163 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00010984
EINECS: 233-039-9

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR