Kobalt(II)nitrat heksahydrat ≥97.7%

Leverandør: Thermo Fisher Scientific
Danger

11341.14 11341.30 11341.A1 11341.A7
11341.14EA 925 NOK
11341.14 11341.30 11341.A1 11341.A7
Kobalt(II)nitrat heksahydrat ≥97.7%
Kobalt(II)nitrat heksahydrat

Purity: 97.7% min CAS No.: 10026-22-9 Molecular Formula: Co(NO3)2.6H2O Formula Weight: 291.04 (182.94anhy) Physical Form: Crystalline Density: 1.87 Refractive Index: 1.52 Appearance: Red Odour: Odourless Melting Point: 55-56° Merck Reference: 14,2444 UN No.: 1477 MDL No.: MFCD00149647

Formel: Co(NO₃)₂·6H₂O
Smeltepunkt: 55…56 °C
Tetthet: 1,87
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149647
CAS-nummer: 10026-22-9
EINECS: 233-402-1
FN: 1477
ADR: 5.1,III
Merck Indeks: 14,2444

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR