Kadmium ≥99.9975% (metals basis), foil, Puratronic®, Thickness: 0.25 mm (0.01 in)

Leverandør: Thermo Fisher Scientific
Danger

11370.FIEA 615 NOK
11370.FI 11370.GH
Kadmium ≥99.9975% (metals basis), foil, Puratronic®, Thickness: 0.25 mm (0.01 in)
Kadmium

Purity: 99.9975% CAS No.: 7440-43-9 Physical Form: ≈5.4g/50x50mm MDL No.: MFCD00010914

Formel: Cd
MW: 112.41 g/mol
Kokepunkt: 765
Smeltepunkt: 321
Tetthet: 8.642 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00010914
CAS-nummer: 7440-43-9
EINECS: 231-152-8
FN: 2570
ADR: 6.1 II
Merck Indeks: 12,01649

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR