ESD safety shoes, lace-up, uvex 2, vibram®, 6522
Leverandør: UVEX
6522.2/43 6522.2/41 6522.2/42 6522.2/45 6522.2/49 6522.2/50 6522.2/47 6522.2/48 6522.2/51 6522.2/52 6522.2/44 6522.2/46 6522.2/37 6522.2/38 6522.2/35 6522.2/36 6522.2/39 6522.2/40 6522.4/36 6522.4/37 6522.4/38 6522.4/39 6522.3/35 6522.3/36 6522.3/37 6522.3/38 6522.3/39 6522.3/40 6522.3/41 6522.3/42 6522.3/43 6522.3/44 6522.3/45 6522.3/46 6522.3/47 6522.3/48 6522.3/49 6522.3/50 6522.3/51 6522.3/52 6522.4/45 6522.4/46 6522.4/35 6522.4/40 6522.4/41 6522.4/42 6522.4/43 6522.4/44 6522.4/47 6522.4/48 6522.4/49 6522.4/50 6522.4/51 6522.4/52
114-8067EA 2175 NOK
114-8067 114-8065 114-8066 114-8069 114-8073 114-8074 114-8071 114-8072 114-8075 114-8076 114-8068 114-8070 114-8061 114-8062 114-8059 114-8060 114-8063 114-8064 114-8667 114-8668 114-8669 114-8670 114-8648 114-8649 114-8650 114-8651 114-8652 114-8653 114-8654 114-8655 114-8656 114-8657 114-8658 114-8659 114-8660 114-8661 114-8662 114-8663 114-8664 114-8665 114-8640 114-8641 114-8666 114-8635 114-8636 114-8637 114-8638 114-8639 114-8642 114-8643 114-8644 114-8645 114-8646 114-8647
ESD safety shoes, lace-up, uvex 2, vibram®, 6522
Sko og støvler
Metal-free, lightweight, flexible and robust safety shoe made from breathable, water repellent high-tech microvelour upper, soft padded tongue and collar, and a dual-density slip-resistant VIBRAM® rubber sole resistant to chemicals, oils and gasoline.
  • Climazone technology for improved comfort
  • Breathable distance mesh lining
  • Removable anti-static comfort insole with moisture control system; orthopaedic insoles possible
  • Virtually seam-free construction to eliminate pressure points
  • Lightweight two-layer sole with excellent shock absorption outstanding slip