ISO AIR cleanroom masks with duckbill
Leverandør: Medicom
60.754L 60.754M
114-9496EA 2625 NOK
114-9496 114-9495
ISO AIR cleanroom masks with duckbill
Masker Munnbind, renrom
ISO AIR cleanroom masks with duckbill feature a large and comfortable breathing chamber.
  • Made in France
  • Made from 30% recycled polypropylene material
  • High breathability
  • ISO 3 Grade B protection
Masks feature low particle release, and are Helmke drum tested 'excellent' for filtration efficiency (BFE, PFE, VFE >99%). Softex technology limits the risk of particulate contamination while reducing the risk of skin irritation. An adjustable and malleable nose clip prevents glasses from fogging up. Duckbill-style masks feature a large and comfortableMore Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Sustainable Materials,Sustainable Packaging
Miljøsertifikat Ecovadis, FSC