Cleanroom Earloop Facemasks with Clips, Sterile, Exa 230
Leverandør: ISOFIELD IRELAND
60011
114-9593EA 7550 NOK
114-9593
Cleanroom Earloop Facemasks with Clips, Sterile, Exa 230
Masker Munnbind, renrom
The Isofield Exa is the widest validated sterile facemask available.
  • ISO Class 4, GMP Grade A compatible
  • Extra wide at 230 mm
  • Earloops with clip
  • Validated sterile
  • No-smell gamma compatible materials
  • Excellent BFE and PFE
  • Malleable alluminium nose strip
Measuring 230 mm wide, with comfortable ear loops, the Exa is also fitted with a convenient clip to adapt to your SOP, and is made with gamma compatible materials to ensure low/no odour in use. These individually packed Exa facemasks provide excellent bacterial and particle filtration efficiency, andMore Product Information