Tie-On Cleanroom Facemasks, Sterile, Tera 230
Leverandør: ISOFIELD IRELAND
60131
114-9594EA 7100 NOK
114-9594
Tie-On Cleanroom Facemasks, Sterile, Tera 230
Masker Munnbind, renrom
The Isofield Tera is the widest validated sterile facemask available.
  • ISO Class 4, GMP Grade A compatible
  • Four ties made from polyester
  • Validated sterile
  • No-smell gamma compatible materials
  • Excellent BFE and PFE
  • Malleable alluminium nose strip
Measuring 230 mm wide, with comfortable polyester ties, the Tera is made from gamma compatible materials ensuring low/no odour in use. The individually packed Tera facemasks provide excellent bacterial and particle filtration efficiency, and are GMP Grade A compatible.
Forpakning: Each piece is sealed in HDPEMore Product Information