Cleanroom Earloop Facemasks with Clips, Non Sterile, Peta 230
Leverandør: ISOFIELD IRELAND
60210
114-9595EA 6150 NOK
114-9595
Cleanroom Earloop Facemasks with Clips, Non Sterile, Peta 230
Masker Munnbind, renrom
The Isofield Peta facemask is ISO Class 4 compatible.
  • ISO Class 4 compatible
  • Extra wide at 230 mm
  • Earloops with clip
  • Individually packed
  • Low CFU
  • Excellent BFE and PFE
  • Malleable alluminium nose strip
Measuring 230 mm wide, with comfortable ear loops, the Peta is also fitted with a convenient clip. The individually packed Peta facemasks provide excellent bacterial and particle filtration efficiency.
Forpakning: Each piece is sealed in HDPE bag. 50 sealed individual bags per inner bag, 12 inner bags per case (600 pieces).