Tie-On Cleanroom Facemasks, Non Sterile, Zetta 230
Leverandør: ISOFIELD IRELAND
60330
114-9596EA 5675 NOK
114-9596
Tie-On Cleanroom Facemasks, Non Sterile, Zetta 230
Masker Munnbind, renrom
The Isofield Zetta facemask is ISO Class 4 compatible.
  • ISO Class 4 compatible
  • Extra wide at 230 mm
  • Four ties made from polyester
  • Individually packed
  • Low CFU
  • Excellent BFE and PFE
  • Malleable alluminium nose strip
Measuring 230 mm wide, with comfortable polyester ties, the Zetta is made from low-linting cleanroom grade material to ensure best in class cleanliness. The individually packed Zetta facemasks provide excellent bacterial and particle filtration efficiency.
Forpakning: Each piece is sealed in HDPE bag. 50 sealed individual bags per inner bag,More Product Information