Cobalt(III) fluoride ≥99%

Leverandør: Thermo Fisher Scientific
Danger

Synonymer: Cobaltic fluoride, Cobalt trifluoride

11490.14 11490.22
11490.14EA 1375 NOK
11490.14 11490.22
Cobalt(III) fluoride ≥99%
Cobalt(III) fluoride

Purity: 99% CAS No.: 10026-18-3 Molecular Formula: CoF3 Formula Weight: 115.93 Physical Form: Powder Density: 3.88 Merck Reference: 14,2430 UN No.: 3085 MDL No.: MFCD00010942

Formel: CoF₃
MW: 115.93 g/mol
Smeltepunkt: 927
Tetthet: 3.88
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00010942
CAS-nummer: 10026-18-3
EINECS: 233-062-4
FN: 3085
ADR: 5.1 III
Merck Indeks: 12,02491

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR