Antimony(III) fluoride ≥99%

Leverandør: Thermo Fisher Scientific
Danger

11492.22 11492.36
11492.22EA 945 NOK
11492.22 11492.36
Antimony(III) fluoride ≥99%
Antimony(III) fluoride

Purity: 99+% CAS No.: 7783-56-4 Molecular Formula: SbF3 Formula Weight: 178.75 Physical Form: Crystalline Density: 4.379 Melting Point: 292° Boiling Point: 376° Merck Reference: 14,709 UN No.: 2923 MDL No.: MFCD00011218

Formel: F₃Sb
Kokepunkt: 376 °C
Smeltepunkt: 292 °C
Tetthet: 4,379
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011218
CAS-nummer: 7783-56-4
EINECS: 232-009-2
FN: 2923
ADR: 8,III
Merck Indeks: 14,709

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR