VWR®, Wipes, Pre-Saturated
Leverandør: VWR Collection
115-0128EA 615 NOK
115-0128
VWR®, Wipes, Pre-Saturated
Papirservietter Tørkepapir og våtservietter Våtservietter
These non woven wipes are constructed from 100% polypropylene and are moistened with a blend of pure, reagent grade 70% isopropyl alcohol and 30% reagent grade deionised water. They are ideal for wiping optical parts, delicate glassware, electrodes, cuvettes, microscopes or lenses.
  • Suitable for all applications in environmentally-sensitive areas
  • Very strong, soft and lint-free
  • Will not scratch surfaces
  • Individually dispensed wipes are handy and ready for instant use
  • Reclosable bench top container ensures a clean wipe every time, snap-on sealed top