Cleanroom wipes, pre-saturated, PROSAT® PS-919

Leverandør: CONTEC
PS-919
115-0377EA 360 NOK
115-0377
Cleanroom wipes, pre-saturated, PROSAT® PS-919
Papirservietter Renromsservietter Våtservietter, renrom
This non woven, meltblown PP wipe is pre-saturated with 91% isopropanol and 9% deionised water.

  • Designed for the semiconductor industry
  • Meltblown polypropylene provides improved durability
  • Solvent blend meets SEMI specifications for metals and contaminants
  • Pouches preserve cleanliness and solvent saturation level

Ideal for cleaning process tools and equipment as well as for wiping articles prior to pass through.

Suitable for use in Class 100, ISO 5, M3,5 cleanrooms.

Colour: White

NB: *Produktet/produktene på denne siden er utenfor omfangetMore Product Information
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR