Cleanroom wipes, Anticon® Milliester® Polyester
Leverandør: CONTEC
495352-853
115-0445EA 820 NOK
115-0445
Cleanroom wipes, Anticon® Milliester® Polyester
Papirservietter Renromsservietter Tørkepapir, renrom
Made from high quality, unlaundered 100% continuous filament polyester.
  • Low-linting wipes with excellent absorption properties for use in less critical applications
  • Ideal for cleaning that requires tear- and abrasion-resistance; suitable for hybrid electronics
Suitable for use in Class 10 000, ISO Class 7, M5.5 environments.
Farge: hvit