Wipes, optical grade, Amplitude™ Optic Wipe
Leverandør: CONTEC
C6-99
115-0579EA 635 NOK
115-0579
Wipes, optical grade, Amplitude™ Optic Wipe
Papirservietter Renromsservietter Tørkepapir, renrom
These absorbent non abrasive wipes consist of long staple cellulosic fibres which are wet laid in a paper making process to form an extremely clean sheet.
  • Non woven, long staple fibre construction
  • Very low extractables level, moderate particle generation
  • Can be used as separating layers between CDs and silicon wafers and for packaging and storage of optical products
Ideal for delicate cleaning of optical products, drying critical products and for use as a lining for cleanroom surfaces or trays.
Colour: White.
NB: Data accurate at time ofMore Product Information