Hand disinfectants, Septigel+
Leverandør: LABORATOIRES PRODENE KLINT
B1587PA800Q
115-0627EA 1175 NOK
115-0627
Hand disinfectants, Septigel+
Hudpleie Hånddesinfeksjon Liquid and Foam Skin Disinfectants
Septigel+ is a skin-friendly, moisturising and fast acting hand disinfectant for everyday use.
  • Bactericidal: Active on Salmonella enterica, Listeria monocytogenes
  • Virucidal: Active on HIV, Rotavirus, Herpes virus, BVDV (Hepatitis C indicator), PRV (Hepatitis B indicator), MNV (murine norovirus, model virus for the study of human noroviruses), Poliovirus, Adenovirus
  • Fungicidal: Candida albicans, Aspergillus niger
  • Tuberculocidal: Mycobacterium terrae
  • Contains ethanol, isopropanol, glycerine
  • No rinsing required
  • Colourant- and perfume-free
  • Hypoallergenic