Cleaning cloths, Tork Industrial
Leverandør: Essity
520337 520304
115-0638EA 1275 NOK
115-0638 115-3122
Cleaning cloths, Tork Industrial
Papirservietter Rengjøringskluter for gjenbruk Kluter til generell bruk
These cleaning cloths made from virgin cellulose pulp, polyester fibres and PP fibres are a perfect alternative to rags or rental cloths.
  • exelCLEAN® technology – for a quicker professional cleaning result
  • Strong texture for excellent oil and water absorption
  • Meet all cleaning needs due to their suitability with most solvents and detergents
  • Tork Easy Handling™ for easier carrying, opening and disposing of packaging
  • Silicone-free: Ensures that there are no residues on a surface
  • Suitable for different dispensing systems
Their softness and flexibilityMore Product Information