Cleanroom wipes, pre-saturated, PROSAT® PS-SI
Leverandør: CONTEC
PS-SI
115-0911EA 20425 NOK
115-0911
Cleanroom wipes, pre-saturated, PROSAT® PS-SI
Papirservietter Renromsservietter Våtservietter, renrom
This durable wipe is made from 100% non woven polyester fibers, hydroentagled into an extremely soft, clean, and strong fabric which is very low in both particles and extractables.
  • Solution blend of 10% 2-propanol and 90% deionized water combines cleaning effectiveness with safety
  • Semiconductor grade solution is 0,2 µm filtered and meets SEMI specifications for metals
  • Low IPA concentration for increased safety in semiconductor facilities
  • Available in convenient, easy to use peel/reseal pouches
Designed for the semiconductor industry.
Suitable forMore Product Information