Hand cleansing wipes, pre-saturated, Kresto kwik-wipes®
Leverandør: Deb-STOKO
23122
115-1473EA 1650 NOK
115-1473
Hand cleansing wipes, pre-saturated, Kresto kwik-wipes®
Papirservietter Tørkepapir og våtservietter Våtservietter
These polypropylene wipes are moistened with a cleansing solution that is ideal for fast cleansing and pre-cleansing after contact with heavy-duty contamination, such as waste mineral and lubricating oils, graphite, metallic dust and carbon black.
  • Ideal for workplaces where there is no water available for cleaning hands
  • Slightly roughened surface provides consistent and reliable cleansing power
  • Non soapy wipes help to stop skin swelling
  • Slightly acidic pH helps to maintain the natural acidic protective mantle
NB: *Produktet/produktene på denne sidenMore Product Information