Surface preparation cloth, KIMTECH PREP*
Leverandør: KIMBERLY CLARK
7589
115-1554EA 5475 NOK
115-1554
Surface preparation cloth, KIMTECH PREP*
Papirservietter Rengjøringskluter for gjenbruk Kluter til generell bruk
This highly absorbent multipurpose cleaning and polishing cloth made from microfibre can be used wet or dry. It is effective in trapping and removing particulate matter from a variety of surfaces.
  • Low linting which reduces the risk of process contamination
  • Machine washable up to 300 times
  • Soft and flexible
  • Silicone-free, seamed edges