Sealent wipes, KIMTECH PREP*
Leverandør: KIMBERLY CLARK
7642
115-1556EA 7975 NOK
115-1556
Sealent wipes, KIMTECH PREP*
Papirservietter Tørkepapir og våtservietter Papirservietter, skånsom
These soft and flexible wipes have a non woven open structure that is ideal for the removal of excess sealant prior to industrial painting or coating of a surface.
  • Textured open weave material traps and removes excess sealant
  • Pliable material helps remove sealant from hard to get to spaces
  • Low linting performance helps minimise the occurrence of defects and cross-contamination from particulate matter