VWR® B'Wipes 6000, Cleanroom wipes
Leverandør: VWR Collection
115-1821EA 1050 NOK
115-1821 115-1823
VWR® B'Wipes 6000, Cleanroom wipes
Papirservietter Renromsservietter Tørkepapir, renrom
2 ply polyester sealed-border wipe for use in ISO 3 cleanrooms.
  • Quilted polyester wipe, constructed from 2 plies, with sealed borders
  • Laundered, with sealed borders, for excellent cleanliness
  • 2 ply quilted, for extreme absorption
  • Ideal for spill control in critical areas, and for applying a cleaning/disinfecting solution over a large surface