VWR® B'Wipes 6000E, Cleanroom floor wipes
Leverandør: VWR Collection
115-1830EA 1300 NOK
115-1830
VWR® B'Wipes 6000E, Cleanroom floor wipes
Papirservietter Renromsservietter Tørkepapir, renrom
Double ply polyester sealed-edge wipe for use in ISO 5 cleanrooms.
  • Economical version of the series 6000, especially for floor cleaning
  • Two plies quilted for superior absorption
  • Ideal for applying a large quantity of cleaning/disinfecting solution on the floors, maximising the number of square metres covered with a single wipe