Cleanroom wipes, pre-saturated, PROSAT® Polynit Heatseal
Leverandør: CONTEC
PSPS0046
115-1869EA 22025 NOK
115-1869
Cleanroom wipes, pre-saturated, PROSAT® Polynit Heatseal
Papirservietter Renromsservietter Våtservietter, renrom
This wipe is made from filament polyester that is laser cut to bond the fibres at the edges of the wipe. This creates a very clean, non abrasive edge that allows full utilisation of the wipe surface. Pre-saturated with 5% semi grade IPA and 95% DIW.
  • Wipe is exceptionally low in particles and extractable residue
  • Presaturation greatly reduces solvent usage and maximises process control and consistency
  • Abrasion and chemical resistant
  • Ideal for general purpose and critical cleaning
Farge: Hvit
NB: Data accurate at time of publishing, latest testMore Product Information