Canister for cleanroom wipes, pre-saturated, SATWipes®
Leverandør: CONTEC
SAT-CAN120
115-1874EA 2150 NOK
115-1874
Canister for cleanroom wipes, pre-saturated, SATWipes®
Dispensere for tørkekluter
This reusable canister holds pre-saturated wipes that are 6" x 9" (152×229 mm) and are dispensed from center-pull rolls.
Canister comes with a snap close lid that helps keep wipes saturated and clean.
NB: *Produktet/produktene på denne siden er utenfor omfanget av EU -direktivet 2019/904