Wipes, KIMTECH PREP* WETTASK*
Leverandør: KIMBERLY CLARK
7762
115-1997EA 10650 NOK
115-1997
Wipes, KIMTECH PREP* WETTASK*
Papirservietter Tørkepapir og våtservietter Våtservietter
This refillable wet wiping system is the ideal alternative to using a spray bottle or open bucket in combination with a rag, making it easy to use and versatile. Two different types, depending on application.
  • Closed-bucket system keeps the pre-moistened wipes fresh in the container and avoids contamination
  • Designed to be compatible with quaternary amine disinfectants
  • Reduces exposure to chemical vapours, splashes and spills
SXX for working with solvents, very strong and absorbent rayon/polyester, no loose fibres. Solvent resistant won't break downMore Product Information