Protective barrier cream, Stokoderm® Universal PURE
Leverandør: Deb-STOKO
34778
115-2250EA 6600 NOK
115-2250
Protective barrier cream, Stokoderm® Universal PURE
Hudpleie Kremer Hand Creams
Triple action cream protects against water and oil based substances, plus helps improve grip and comfort under gloves. Perfume-free and dye-free.
  • Unique triple action formulation
  • Contains Zinc Salicylate astringent agent and a blend of skin caring ingredients
  • Proven general purpose protection and glove compatibility
  • Formulated with a low pH value (4.7-5.2)
  • Hypoallergenic
  • Silicone-free
Suitable for most workplaces to protect against oil-based and water-based substances (such as oil, grease, lubricants, and detergents) and where gloves are worn. Apply 1More Product Information