VWR®, Industrial Wipes, Melt-Blown
Leverandør: VWR Collection
115-2418EA 1575 NOK
115-2418
VWR®, Industrial Wipes, Melt-Blown
Papirservietter Tørkepapir og våtservietter Papirservietter til generell bruk
These melt-blown wipes, made from 100% PP, have a very high absorption capacity.
  • Absorb water, grease and oil
  • Resistant to acids and solvents
  • Soft, synthetic and colour-fast
  • Recyclable
  • Silicone-free
  • Food contact certified
Typical applications include: Aerospace, automotive, electronics, food and beverage, laboratories, printing and janitorial tasks.
Basis weight: 70 g/m²
Absorption: 660% (water); 1350% (oil)