Multi-layer adhesive floor mats, Nomad™ 4300
Leverandør: 3M
ADHWS60
115-2706EA 26200 NOK
115-2706
Multi-layer adhesive floor mats, Nomad™ 4300
Gulvmatter Renromsmatter
4300 contamination control mats consist of 40 layers of adhesive-coated clear polyethylene sheets attached to a vinyl backing. For use at cleanroom entrances, hospitals, semi-conductor, medical device, pharmaceutical, biological and nuclear industries, also at factory to office entrances and other work areas requiring high levels of cleanliness.
  • Backing adheres to the floor, while the tacky top surface removes dirt and contaminants from the bottom of shoes and wheels
  • Very flat; no frame needed
Colour: Blue or white